Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cerium(Iv) Sulfate Tetrahydrate, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
Specifications
Quantity | 100 g |
Linear Formula | Ce(SO4)2 · 4H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 404.3g/mol |
Percent Purity | ≥98% |
Grade | puriss. p.a. |
Chemical Name or Material | Cerium(Iv) Sulfate Tetrahydrate |
Safety and Handling
P305 + P351 + P338
H315-H319
EINECSNumber : 236-644-6