Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cerium sputtering target, 50.8mm dia x 3.18mm thick

Manufacturer:  Alfa Aesar™ 45734KS

Quantity 1 ea.

 View more versions of this product

Catalog No. AA45734KS


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-45-1
MFCD00010924
UN1333
Cerium sputtering target, 50.8mm dia. x 3.18mm thick

Specifications

1 ea.
Odorless
UN1333
7440-45-1
MFCD00010924
Cerium sputtering target, 50.8mm dia. x 3.18mm thick

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit