Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cerium tungsten oxide, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 3584014

Quantity 25g

 View more versions of this product

Catalog No. AA3584014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13454-74-5
MFCD00049725
Cerium tungsten oxide
1023.78
Cerium tungstate

Specifications

13454-74-5
1023.78
Cerium tungstate
Odorless
25g
99.9%, (REO)
Powder
MFCD00049725
−100 Mesh
1089°C
Cerium tungsten oxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit