Alfa Aesar™ Cerium vanadium oxide, 99.9% (REO)

Manufacturer: Alfa Aesar™ 4020914

Quantity 25g

 View more versions of this product

Catalog No. AA4020914

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13597-19-8
    Formula Weight 255.06
    MDL Number MFCD00171262
    UN Number UN3285
    Synonym Cerium vanadate
    Chemical Name or Material Cerium vanadium oxide

    Specifications

    Formula Weight 255.06
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13597-19-8
    Health Hazard 2 GHS H Statement
    H331-H302-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder
    MDL Number MFCD00171262
    UN Number UN3285
    Synonym Cerium vanadate
    Quantity 25g
    Health Hazard 1 Danger
    TSCA TSCA
    Mesh Size −200 Mesh
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
    Chemical Name or Material Cerium vanadium oxide
    Assay 99.9%, (REO)

    GHS H Statement
    H331-H302-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit