Certified Safety Aspirin Pain Relief


 View all options for this product

Catalog No. 19-075369

  Check Availability
Add to Cart


    Quantity 200 Case
    Product Type Aspirin Tablets
    Active Ingredients Aspirin 325mg
    Format Tablet
    For Use With (Application) Provides temporary relief of minor aches, pains and fever

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit