Certified Safety Aspirin Pain Relief

Manufacturer: CERTIFIED SAFETY MANUFACTURING 232014

 View more versions of this product

Catalog No. 19-808-944

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Active Ingredients Aspirin 325mg
    For Use With (Application) Provides temporary relief of minor aches, pains and fever
    Format Tablet
    Quantity 500 Pack
    Product Type Aspirin Tablets

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit