Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Certified Safety Certi-Cet Plus Acetaminophen Pain Relief

Specifications

Product Type Certi-Cet Plus Acetaminophen Tablet
Format Tablet
For Use With (Application) Provide relief from headache, sprains and fever
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
19-808-947
View Documents
Certified Safety
232102
500 Pack This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

Certi-Cet Plus Acetaminophen Tablet
Tablet
Provide relief from headache, sprains and fever
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit