Certified Safety Certi-Cet Plus Acetaminophen Pain Relief

Manufacturer: CERTIFIED SAFETY MANUFACTURING 232104

 View more versions of this product

Catalog No. 19-041-952

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Active Ingredients Acetaminophen 325mg
    For Use With (Application) Provide relief from headache, sprains and fever
    Format Tablet
    Quantity 12 Case
    Product Type Certi-Cet Plus Acetaminophen Tablet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit