Learn More
Certified Safety Certified Safety Certi-Ose Eye Washes
Isotonic buffered solution provides safe, effective washing
Manufacturer:  Certified Safety R511017
 View more versions of this product
Catalog No. 19-120-313
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Ideal for flushing small objects out of the eye
  • Safe for use on skin
  • Sterile
  • Twist-off top
  • Volume: 20mL
Specifications
Specifications
0.67 oz.
19.81mL
0.67 oz., 19.81mL
Eyewash