Certified Safety CPRotector™ Mini-Pouch

Protection always within reach.

Manufacturer: CERTIFIED SAFETY MANUFACTURING 270037

 View more versions of this product

Catalog No. 19-808-986

  Check Availability
Add to Cart

    Specifications

    Color Orange
    Disposable Yes
    Includes One CPRotector and one pair of Nitrile gloves, antiseptic towelette
    For Use With (Application) Prevents unsafe mouth-to-mouth contact
    Product Type CPRotector mini pouch kit with gloves and antiseptic towelette

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit