Certified Safety LXXV and CLX First Aid Kits

Manufacturer: CERTIFIED SAFETY MANUFACTURING K206140

 View more versions of this product

Catalog No. 19-035-117

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Depth (English) 5.5 in.
    Depth (Metric) 13.97cm
    Height (English) 16 in.
    Includes Plastic adhesive bandages, woven adhesive bandages, tri-cut adhesive tape, alcohol wipes, antiseptic spray, antiseptic towelettes, aspirin tablets, biohazard bags, burn spray, cold packs, cotton, cotton swabs, CPRotector 2000™ mouth-to-m
    Width (English) 6 in.
    Product Type CLX - 100 Person - Deluxe First aid Kit
    Certifications/Compliance ANSI Z308.1-1998
    Height (Metric) 40.64cm
    Width (Metric) 13.4cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit