Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Certified Safety Unitized First Aid Kits
SDP

Kits stay neat and organized, fitting snugly to prevent shifting.

Specifications

Product Type First Aid Kit
Height (English) 5 in.
Certifications/Compliance ANSI
Includes Handle and bracket for wall mounting, certi-strips - Plastic , certi-strips - Plastic - extra large, certi-gauze bandage compress, triangular bandage with Pins, certi-gauze pad, certi-strips - heavy weight woven, absorbent compress, Gloves - Nitrile, certi-sporyn - antibiotic cream, Antiseptic BZK towelettes, PVP iodine wipe-ups, certi-tape, cold pack - certi-cool junior, certi-burn cream, eye wash, alcohol gel, first aid facts guide
Height (Metric) 12.7 cm
View More Specs

Products 2
Catalog Number Mfr. No. Depth (Metric) Product Type Height (Metric) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Depth (Metric) Product Type Height (Metric) Width (Metric) Price Quantity & Availability  
19-808-823
SDP
View Documents
Certified Safety
K202012
7.62 cm First Aid Kit 12.7 cm 24 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-808-824
SDP
View Documents
Certified Safety
202014
6.35 cm First Aid Kit 12.7 cm 10.16 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Single-use packaging
  • List of contents permanently mounted inside the lid
  • Dust- and moisture-resistant
  • Open lid of steel kits can be used as shelf when mounted on wall
  • Glow in the dark

Compliance

Meet ANSI™ standards

Specifications

Specifications

First Aid Kit
ANSI
12.7 cm
5 in.
Handle and bracket for wall mounting, certi-strips - Plastic , certi-strips - Plastic - extra large, certi-gauze bandage compress, triangular bandage with Pins, certi-gauze pad, certi-strips - heavy weight woven, absorbent compress, Gloves - Nitrile, certi-sporyn - antibiotic cream, Antiseptic BZK towelettes, PVP iodine wipe-ups, certi-tape, cold pack - certi-cool junior, certi-burn cream, eye wash, alcohol gel, first aid facts guide
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit