Learn More
Aviva Systems Biology Corp CES7 Antibody - N-terminal region (ARP51162_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CES7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CES7. Alternate names: CAUXIN, CES4C1, CES5, FLJ31547, CES7ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60505