Learn More
CES7 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 195201AP
Catalog No. 50-173-2431
Add to cart
Description
Description
The antibody can recognize the isoform1 and isoform2.
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.
Specifications
Specifications
CES7
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6NT32
Carboxylesterase 7, CAUXIN, CES4C1, CES5, CES7
Rabbit
IgG
150 μL
-20°C
Primary
221223
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CES5A
Liquid
CES5A
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human