Cesium, 99.98% (metals basis), Alfa Aesar

Manufacturer: Alfa Aesar 1014622

Quantity 100g

 View more versions of this product

Catalog No. AA1014622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-46-2
    Physical Form Solid
    Assay Percent Range 99.98% (Metals basis)
    Molecular Formula Cs
    Formula Weight 132.9054
    MDL Number MFCD00134037
    Chemical Name or Material Cesium

    Specifications

    Formula Weight 132.9054
    Quantity 100g
    Color Silver
    DOT Information DOT Class: 4.3, Packaging Group: I
    EINECS Number 231-155-4
    Physical Form Solid
    Assay Percent Range 99.98% (Metals basis)
    Molecular Formula Cs
    MDL Number MFCD00134037
    CAS 7440-46-2
    Odor Odorless
    Appearance Silver-colored solid
    Vapor Pressure 0.0000011 hPa
    Health Hazard 1 H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Cesium

    H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit