Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium bicarbonate, 99.9%, pure, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
15519-28-5
CHCsO3
193.921
MFCD00010961
ZMCUDHNSHCRDBT-UHFFFAOYSA-M
cesium bicarbonate,cesium hydrogen carbonate,caesium bicarbonate,caesium 1+ ion bicarbonate,cesium hydrogencarbonate,acmc-1cmhe,cho3.cs,caesium 1+ bicarbonate,carbonic acid, cesiumsalt 1:1
2734983
cesium;hydrogen carbonate
C(=O)(O)[O-].[Cs+]
15519-28-5
CHCsO3
193.921
MFCD00010961
ZMCUDHNSHCRDBT-UHFFFAOYSA-M
cesium bicarbonate,cesium hydrogen carbonate,caesium bicarbonate,caesium 1+ ion bicarbonate,cesium hydrogencarbonate,acmc-1cmhe,cho3.cs,caesium 1+ bicarbonate,carbonic acid, cesiumsalt 1:1
2734983
cesium;hydrogen carbonate
C(=O)(O)[O-].[Cs+]
Assay Percent Range 99.9%
Linear Formula CsHCO3
Fieser 08,81
Formula Weight 193.92
Physical Form Adhering Crystalline Powder or Crystals and Chunks
Grade Pure
Packaging Glass bottle
Total Trace Metal Impurities 0.1% max.
Color White
Quantity 100g
Chemical Name or Material Cesium bicarbonate, 99.9%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 239-554-5