Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium bromide, 99.9%, pure, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
7787-69-1
BrCs
212.81
MFCD00010954
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium bromide,caesium bromide,cesium bromide csbr,tricesium tribromide,csbr,unii-06m25edm3f,caesium 1+ ion bromide,cesiumbromide,br.cs,caesium i bromide
24592
cesium;bromide
[Br-].[Cs+]
7787-69-1
BrCs
212.81
MFCD00010954
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium bromide,caesium bromide,cesium bromide csbr,tricesium tribromide,csbr,unii-06m25edm3f,caesium 1+ ion bromide,cesiumbromide,br.cs,caesium i bromide
24592
cesium;bromide
[Br-].[Cs+]
Assay Percent Range 99.90%
Linear Formula CsBr
Merck Index 15, 2008
Solubility Information Solubility in water: 55g/100mL (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone
Formula Weight 212.81
Physical Form Crystals or Crystalline Powder
Percent Purity 99.9%
Grade Pure
Density 4.4400g/mL
Packaging Glass bottle
Solubility Solubility in water: 55g/100mL (20 c). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Specific Gravity 4.44
Total Trace Metal Impurities 0.1% max.
Boiling Point 1300.0°C
Color White
Melting Point 636.0°C
Quantity 100g
Chemical Name or Material Cesium bromide, 99.9%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 232-130-0

RTECSNumber : FK9275000

TSCA : TSCA