Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Carbonate, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
534-17-8
534-17-8
Grade Reagent
CAS Max % 1
Percent Purity 0.98
Packaging Poly Bottle
Quantity 100 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.