Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407366
Click to view available options
Quantity:
10 g
50 g
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
caesium(1+) chloride
[Cl-].[Cs+]
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
caesium(1+) chloride
[Cl-].[Cs+]

Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds an application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1303°C
Melting Point 646°C
Color White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99.99%
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
Formula Weight 168.36
Odor Odorless
Refractive Index 1.6418
Sensitivity Hygroscopic
Appearance White powder/crystalline/beads
Density 3.99 g/cm3 at 20°C
Chemical Name or Material Cesium chloride
Show More Show Less
Hazard Category H361-H373
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 231-600-2
RTECSNumber FK9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit