Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
For ultra trace analysis, For UV/VIS spectroscopy
Absorbance (50% aqueous solution) 1 cm layer, (50% aqueous solution) 1 cm layer at 0nm, 0.02 max. at 260nm, 0.02 max. at 280nm
Absorbance Conditions (50%, aqua soln)
Assay Percent Range 99.99%
Linear Formula CsCl
Merck Index 15, 2010
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol,insoluble in acetone
Formula Weight 168.36
Physical Form Liquid
Percent Purity 99.99%
Grade Trace Metal Basis
Insoluble in Water 25ppm max.
Insoluble Matter 25ppm (in water) max
Packaging Glass bottle
Purity 99.94% min. (metals basis)
Solubility Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
Boiling Point 1290.0°C
Color Colorless to Yellow
Melting Point 646.0°C
Quantity 25g
Chemical Name or Material Cesium chloride, For spectroscopy
Show More Show Less

GHS H Statement
Suspected of damaging fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 231-600-2