Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, 99.999%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
7647-17-8
ClCs
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride,caesium chloride,cesium monochloride,cesium chloride cscl,dicesium dichloride,cscl,tricesium trichloride,unii-gnr9hml8ba,gnr9hml8ba,caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
7647-17-8
ClCs
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride,caesium chloride,cesium monochloride,cesium chloride cscl,dicesium dichloride,cscl,tricesium trichloride,unii-gnr9hml8ba,gnr9hml8ba,caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula CsCl
Merck Index 15,201
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol,insoluble in acetone
Formula Weight 168.36
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
Total Trace Metal Impurities 0.001% max
Boiling Point 1290.0°C
Color White
Melting Point 646.0°C
Quantity 10g
Chemical Name or Material Cesium chloride
Show More Show Less

GHS H Statement
Suspected of damaging fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 231-600-2