Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium chloride, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Plastic bottle
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride,caesium chloride,cesium monochloride,cesium chloride cscl,dicesium dichloride,cscl,tricesium trichloride,unii-gnr9hml8ba,gnr9hml8ba,caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride,caesium chloride,cesium monochloride,cesium chloride cscl,dicesium dichloride,cscl,tricesium trichloride,unii-gnr9hml8ba,gnr9hml8ba,caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
Aluminum (Al) 1ppm max.
Assay Percent Range 99% min. (Argentometry)
Linear Formula CsCl
Merck Index 15, 2010
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol,insoluble in acetone
Barium (Ba) 10ppm max.
Formula Weight 168.36
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 10ppm max.
Chromium (Cr) 2ppm max.
Density 3.983
Iron (Fe) 5ppm max.
Lead (Pb) 5ppm max.
Lithium (Li) 1ppm max.
Magnesium (Mg) 5ppm max.
Manganese (Mn) 5ppm max.
Packaging Plastic bottle
Potassium (K) 30ppm max.
Rubidium (Rb) 50ppm max.
Silicon Dioxide 2ppm max.
Sodium (Na) 50ppm max.
Solubility Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
Sulfate 50ppm max
Trace Analysis 10ppm max.
Boiling Point 1290.0°C
Color White
Melting Point 646.0°C
Quantity 50g
Chemical Name or Material Cesium chloride, p.a.
Show More Show Less

GHS H Statement
Suspected of damaging fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 231-600-2

RTECSNumber : FK9625000

TSCA : TSCA