Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, BiotechGrade, 99.9%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
7647-17-8
ClCs
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
[Cl-].[Cs+]
7647-17-8
ClCs
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
[Cl-].[Cs+]

C1252, 7647-17-8, CsCl

Spectrum™ Cesium Chloride, BiotechGrade is a colorless inorganic compound and a solid. It is used in isopynic centrifugation as well as in analytical chemistry as a reagent.
CAS Max % 100%
Appearance Fine, white, crystal
Heavy Metals (as Pb) 0.002%
Percent Purity 99.9%
Packaging Poly Pail
Quantity 5 kg