Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride, Optical Grade, 99%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
7647-17-8
7647-17-8
Grade Optical Grade
CAS Max % 1
Appearance Colorless Crystal
Percent Purity 0.99
Heavy Metals 5e-06
Iron (Fe) 5e-06
Packaging Amber Glass Bottle
Quantity 100 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.