Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cesium chloride, ultra dry, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4283214

Quantity 25g

 View more versions of this product

Catalog No. AA4283214


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7647-17-8
168.36
14,2011
CsCl
MFCD00010955
Cesium chloride, Ultra dry

Specifications

7647-17-8
CsCl
MFCD00010955
3.99g/mL
Ampule
Hygroscopic
646°C
Cesium chloride, Ultra dry
Powder
168.36
14,2011
Odorless
1.6418
1303°C
25g
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

RTECSNumber : FK9625000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit