Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cesium chromate, 99+%

Click to view available options
Quantity:
10g
50g
250g
13454-78-9
CrCs2O4
381.803
MFCD00016050
BROHICCPQMHYFY-UHFFFAOYSA-N
cesium chromate,cesium chromate 99.9+%,dicesium dioxido dioxo chromium,dicesium chromate,cesium chromate cs2cro4,chromic acid h2cro4 , dicesium salt,caesium chromate,cro4.2cs,acmc-1bym2,dicaesium 1+ chromate 2-
61613
dicesium;dioxido(dioxo)chromium
[O-][Cr](=O)(=O)[O-].[Cs+].[Cs+]
13454-78-9
CrCs2O4
381.803
MFCD00016050
BROHICCPQMHYFY-UHFFFAOYSA-N
cesium chromate,cesium chromate 99.9+%,dicesium dioxido dioxo chromium,dicesium chromate,cesium chromate cs2cro4,chromic acid h2cro4 , dicesium salt,caesium chromate,cro4.2cs,acmc-1bym2,dicaesium 1+ chromate 2-
61613
dicesium;dioxido(dioxo)chromium
[O-][Cr](=O)(=O)[O-].[Cs+].[Cs+]
UN Number UN3087
Formula Weight 381.81
Percent Purity ≥99%
Odor Odorless
Quantity 250g
Chemical Name or Material Cesium chromate

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

TSCA : TSCA