Learn More
Cesium chromate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Cesium chromate is used to produce cesium vapor by reaction with silicon, boron, or titanium, which is used in the final stages of creating vacuum tubes. It is used in the production of photo-tubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN3087 |
Solubility Information | Soluble in water. |
Formula Weight | 381.81 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Assay | (metals basis) |
Chemical Name or Material | Cesium chromate |
Safety and Handling
Hazard Category | H272-H317-H350i |
Hazard Statement | GHS H Statement H350-H317 May cause cancer. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P210-P220-P221-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: II |
EINECSNumber | 236-640-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.