Alfa Aesar™ Cesium chromate, 99+%

Manufacturer: Alfa Aesar™ B2361909

Quantity 10g

 View more versions of this product

Catalog No. AAB2361909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13454-78-9
    Molecular Formula Cs2CrO4
    Formula Weight 381.81
    MDL Number MFCD00016050
    UN Number UN3087
    Synonym Chromic acid cesium salt
    Chemical Name or Material Cesium chromate

    Specifications

    Formula Weight 381.81
    TSCA TSCA
    MDL Number MFCD00016050
    UN Number UN3087
    Synonym Chromic acid cesium salt
    Quantity 10g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
    Odor Odorless
    Molecular Formula Cs2CrO4
    CAS 13454-78-9
    Chemical Name or Material Cesium chromate
    Assay ≥99%

    GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit