Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cesium dichromate, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 4023022

Quantity 100g

 View more versions of this product

Catalog No. AA4023022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13530-67-1
481.8
UN3087
Cs2Cr2O7
MFCD00049436
Cesium dichromate

Specifications

13530-67-1
Cs2Cr2O7
MFCD00049436
Hygroscopic
Cesium dichromate
Coarse Powder
481.8
UN3087
100g
99.8%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire
oxidizer.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit