Alfa Aesar™ Cesium dichromate, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 4023014

Quantity 25g

 View more versions of this product

Catalog No. AA4023014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13530-67-1
    Linear Formula Cs2Cr2O7
    Formula Weight 481.8
    MDL Number MFCD00049436
    UN Number UN3087
    Chemical Name or Material Cesium dichromate

    Specifications

    Formula Weight 481.8
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13530-67-1
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H350-H272-H317
    May cause cancer.
    May intensify fire
    oxidizer.
    May cause an allergic skin reaction.
    Physical Form Coarse Powder
    Sensitivity Hygroscopic
    Linear Formula Cs2Cr2O7
    UN Number UN3087
    MDL Number MFCD00049436
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
    Chemical Name or Material Cesium dichromate
    Assay 99.8%, (metals basis)

    GHS H Statement
    H350-H272-H317
    May cause cancer.
    May intensify fire
    oxidizer.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit