Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium fluoride, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
13400-13-0
CsF
151.91
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride
25953
cesium;fluoride
[F-].[Cs+]
13400-13-0
CsF
151.91
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium fluoride,caesium fluoride,cesium monofluoride,dicesium difluoride,tricesium trifluoride,unii-t76a371hjr,caesium 1+ ion fluoride,caesiumfluoride,cesiumfluoride
25953
cesium;fluoride
[F-].[Cs+]
C-X bond formation reagent
Aluminum (Al) 10ppm max.
Assay Percent Range 98.5% min
Linear Formula CsF
Fieser 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
Merck Index 15,2011
Solubility Information Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol,insoluble in dioxane and pyridine
Barium (Ba) 10ppm max.
Formula Weight 151.91
Physical Form Crystalline Powder
Percent Purity 99%
Grade Analysis
Calcium (Ca) 50ppm max.
Chloride 200ppm max
Chlorine (Cl) 200ppm max.
Lithium (Li) 5ppm max.
Magnesium (Mg) 50ppm max.
Packaging Plastic bottle
Potassium (K) 300ppm max.
Rubidium (Rb) 200ppm max.
Sodium (Na) 100ppm max.
Solubility Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine
Strontium (Sr) 10ppm max.
Sulfate 300ppm max.
Boiling Point 1251.0°C
Color White
Melting Point 682.0°C
Quantity 25g
Chemical Name or Material Cesium fluoride
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging fertility or the unborn child if inhaled.
Contact with a

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISO

GHS Signal Word: Danger

EINECSNumber : 236-487-3