Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Fluoride, Optical Grade, 99%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
13400-13-0
13400-13-0
Grade Optical Grade
CAS Max % 1
Percent Purity 0.99
Packaging Amber Glass Bottle
Quantity 100 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.