Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium formate hydrate, White chunks/powder, 99.9% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1292736

Quantity 500g

 View more versions of this product

Catalog No. AA1292736


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Chunks and Powder
CsOOCH·xH2O
MFCD00039103
99.9% (Metals basis)
177.92
Cesium formate hydrate

Specifications

Chunks and Powder
CsOOCH·xH2O
MFCD00039103
White
Cesium formate hydrate
99.9% (Metals basis)
177.92
Hydrate
500g

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

H315-H319 : Causes skin irritation. Causes serious eye irritation.

EINECSNumber : 222-492-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit