Cesium formate hydrate, White chunks/powder, 99.9% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1292714

Quantity 25g

 View more versions of this product

Catalog No. AA1292714

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Chunks and Powder
    Assay Percent Range 99.9% (Metals basis)
    Molecular Formula CsOOCH·xH2O
    Formula Weight 177.92
    MDL Number MFCD00039103
    Chemical Name or Material Cesium formate hydrate

    Specifications

    Physical Form Chunks and Powder
    Formula Weight 177.92
    Quantity 25g
    Color White
    EINECS Number 222-492-8
    Anhydrous or Hydrate Hydrate
    MDL Number MFCD00039103
    Assay Percent Range 99.9% (Metals basis)
    Molecular Formula CsOOCH·xH2O
    Health Hazard 1 H315-H319 : Causes skin irritation. Causes serious eye irritation.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
    Chemical Name or Material Cesium formate hydrate

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

    H315-H319 : Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit