Alfa Aesar™ Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis)

Manufacturer: Alfa Aesar™ 16505MZ

Quantity 500g

 View more versions of this product

Catalog No. AA16505MZ

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21351-79-1
    Formula Weight 149.91
    MDL Number MFCD00010964
    UN Number UN2681
    Chemical Name or Material Cesium hydroxide

    Specifications

    Formula Weight 149.91
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Air Sensitive
    MDL Number MFCD00010964
    UN Number UN2681
    Quantity 500g
    TSCA TSCA
    Physical Form Liquid
    CAS 21351-79-1
    Health Hazard 1 Danger
    Density 1.725g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CAESIUM HYDROXIDE SOLUTION
    Odor Odorless
    Color Colorless
    Health Hazard 2 GHS H Statement
    H314-H302
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Concentration 50% w/w aq. soln.
    Chemical Name or Material Cesium hydroxide
    Assay 99%, (metals basis)

    GHS H Statement
    H314-H302
    Causes severe skin burns and eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit