Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water, Acros Organics

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
21351-79-1
CsHO
149.91
MFCD00010964
HUCVOHYBFXVBRW-UHFFFAOYSA-M
cesium hydroxide,caesium hydroxide,cesium hydrate,csoh,cesium hydroxide cs oh,cesium hydroxide dimer,cesium hydoxide,cs.ho,caesium 1+ oh,cesium hydroxide solution
62750
CHEBI:33988
cesium;hydroxide
[OH-].[Cs+]
21351-79-1
CsHO
149.91
MFCD00010964
HUCVOHYBFXVBRW-UHFFFAOYSA-M
cesium hydroxide,caesium hydroxide,cesium hydrate,csoh,cesium hydroxide cs oh,cesium hydroxide dimer,cesium hydoxide,cs.ho,caesium 1+ oh,cesium hydroxide solution
62750
CHEBI:33988
cesium;hydroxide
[OH-].[Cs+]
For ultra trace analysis
Aluminum (Al) 5ppm max.
Assay Percent Range 99.9%
Linear Formula CsOH
Merck Index 15, 2012
Solubility Information Solubility in water: soluble
Barium (Ba) 15ppm max.
Formula Weight 149.91
Physical Form Liquid
Percent Purity 99.9%
Grade Trace Metal Basis
Calcium (Ca) 2ppm max.
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Chromium (Cr) 1ppm max.
Density 1.7400g/mL
Iron (Fe) 1ppm max.
Lithium (Li) 1ppm max.
Magnesium (Mg) 1ppm max.
Manganese (Mn) 1ppm max.
Packaging Plastic bottle
Phosphorus Pentoxide 5ppm max.
Potassium (K) 200ppm max.
Rubidium (Rb) 10ppm max.
Silicon Dioxide 50ppm max.
Sodium (Na) 100ppm max.
Solubility Solubility in water: soluble.
Specific Gravity 1.74
Strontium (Sr) 5ppm max.
Sulfate 25ppm max.
Trace Analysis trace analysis values are on dry salt basis
Color Colorless
Quantity 100g
Chemical Name or Material Cesium hydroxide, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 244-344-1

RTECSNumber : FK9800000

TSCA : TSCA