Learn More
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Cesium hydroxide hydrate is used as a catalyst for the conversion of alcohols substituted anilines and heterocyclic amines undergo an addition in NMP to phenylacetylene leading to functionalized enol ethers and enamines. It is also used in the chemoselective N-alkylation of secondary amines with alkyl halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99.9% (Metal basis) |
Solubility Information | Soluble in water and ethanol. |
IUPAC Name | caesium(1+) hydroxide |
Formula Weight | 149.91 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystalline |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335 |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 244-344-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.