Learn More
Cesium hydroxide monohydrate, 96%; cesium carbonate <5%, Thermo Scientific Chemicals
Description
Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.
Solubility
Soluble in water with exothermic reaction.
Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, acids, air, water/moisture.
Specifications
Melting Point | 205°C to 206°C |
Quantity | 5 g |
UN Number | UN3262 |
Merck Index | 14,2013 |
Solubility Information | Soluble in water with exothermic reaction. |
IUPAC Name | cesium;hydroxide;hydrate |
Formula Weight | 167.93 (149.91 Anhydrous) |
Percent Purity | 96% |
Odor | Odorless |
Sensitivity | Air sensitive; Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H302-H314 |
Hazard Statement | GHS H Statement H314-H302 Causes severe skin burns and eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S. |
EINECSNumber | 244-344-1 |
RTECSNumber | FK9800000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.