Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
35103-79-8
HCsO·H2O
167.93
MFCD00149664
ABSOMGPQFXJESQ-UHFFFAOYSA-M
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
23679066
cesium;hydroxide;hydrate
O.[OH-].[Cs+]
35103-79-8
HCsO·H2O
167.93
MFCD00149664
ABSOMGPQFXJESQ-UHFFFAOYSA-M
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
23679066
cesium;hydroxide;hydrate
O.[OH-].[Cs+]
Additional Information Cs2CO3: < 5%
Aluminum (Al) 5ppm max.
Assay Percent Range 99.95%
Linear Formula CsOH·H2O
Merck Index 10, 1970
Solubility Information Solubility in water: soluble with exothermic reaction. Other solubilities: soluble in alcohol (exothermic)
Barium (Ba) 15ppm max.
Formula Weight 167.93
Physical Form Powder
Percent Purity 99.95%
Grade Trace Metal Basis,for analysis
Calcium (Ca) 2ppm max.
Chloride 100ppm max.
Chlorine (Cl) 300ppm max.
Chromium (Cr) 1ppm max.
Iron (Fe) 5ppm max.
Lithium (Li) 1ppm max.
Magnesium (Mg) 1ppm max.
Packaging Plastic bottle
Potassium (K) 150ppm max.
Purity 99.95 % min. (trace metal basis)
Rubidium (Rb) 10ppm max.
Silica 25ppm max.
Silicon Dioxide 30ppm max.
Sodium (Na) 75ppm max.
Solubility Solubility in water: soluble with exothermic reaction. Other solubilities: soluble in alcohol (exothermic)
Strontium (Sr) 5ppm max.
Sulfate 25ppm max.
Trace Analysis 5ppm max.
Color Blue-Red to Green
Melting Point 205.0°C to 208.0°C
Quantity 50g
Chemical Name or Material Cesium hydroxide monohydrate, 99.5%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging fertility or the unborn child.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger