Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium iodide, 99.9%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
7789-17-5
CsI
259.8
MFCD00010962
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
24601
cesium;iodide
[I-].[Cs+]
7789-17-5
CsI
259.8
MFCD00010962
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
24601
cesium;iodide
[I-].[Cs+]
Aluminum (Al) 10ppm max.
Assay Percent Range 99.9%
Linear Formula CsI
Merck Index 15, 2013
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol,slightly soluble in methanol,practically insoluble in acetone
Barium (Ba) 10ppm max.
Formula Weight 259.8
Physical Form Crystalline Powder
Percent Purity 99.9%
Grade Analysis
Calcium (Ca) 10ppm max.
Chromium (Cr) 5ppm max.
Iron (Fe) 10ppm max.
Lithium (Li) 10ppm max.
Magnesium (Mg) 20ppm max.
Manganese (Mn) 5ppm max.
Packaging Glass bottle
Phosphorus Pentoxide 10ppm max.
Potassium (K) 50ppm max.
Purity 99.85% min. (Metal-based)
Rubidium (Rb) 20ppm max.
Silicon Dioxide 10ppm max.
Sodium (Na) 50ppm max.
Solubility Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone
Sulfate 20ppm max.
Trace Analysis 10ppm max.
Boiling Point 1280.0°C
Color White
Melting Point 626.0°C
Quantity 25g
Chemical Name or Material Cesium iodide, 99.9%
Show More Show Less

GHS H Statement
Suspected of damaging fertility or the unborn child if inhaled.
Very toxic to aquatic life.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the

GHS Signal Word: Warning

EINECSNumber : 232-145-2

RTECSNumber : FL0350000

TSCA : TSCA