Alfa Aesar™ Cesium iodide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1002222

Quantity 100g

 View more versions of this product

Catalog No. AA1002222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-17-5
    Formula Weight 259.81
    MDL Number MFCD00010962
    Merck Index 14,2014
    Chemical Name or Material Cesium iodide

    Specifications

    Formula Weight 259.81
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Refractive Index 1.7876
    Sensitivity Hygroscopic
    MDL Number MFCD00010962
    CAS 7789-17-5
    Merck Index 14,2014
    Color White
    Solubility Information Very soluble in water; Soluble in ethanol; Slightly soluble in methanol; Insoluble in acetone
    Health Hazard 1 Warning
    TSCA TSCA
    Mesh Size −20 Mesh
    Boiling Point 1280°C
    RTECS Number FL0350000
    Melting Point 626°C
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H317-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Chemical Name or Material Cesium iodide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H302-H315-H319-H317-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit