Alfa Aesar™ Cesium metavanadate, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3921306

Quantity 5g

 View more versions of this product

Catalog No. AA3921306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14644-55-4
    Formula Weight 231.84
    MDL Number MFCD00049812
    UN Number UN3288
    Synonym Cesium vanadate, meta
    Chemical Name or Material Cesium metavanadate

    Specifications

    Formula Weight 231.84
    Health Hazard 3 GHS P Statement
    P260-P261-P304+P340-P307+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −100 Mesh
    Synonym Cesium vanadate, meta
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H370-H302-H332
    Causes damage to organs.
    Harmful if swallowed.
    Harmful if inhaled.
    Physical Form Powder
    MDL Number MFCD00049812
    Color White to yellow
    Health Hazard 1 Danger
    TSCA TSCA
    Odor Odorless
    CAS 14644-55-4
    UN Number UN3288
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Chemical Name or Material Cesium metavanadate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H370-H302-H332
    Causes damage to organs.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P304+P340-P307+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit