Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium perchlorate 99% , ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass Bottle
13454-84-7
ClCsO4
232.351
MFCD00010956
WKDKOOITVYKILI-UHFFFAOYSA-M
cesium perchlorate,caesium 1+ ion perchlorate,perchloric acid, cesium salt,caesiumperchlorat,caesium perchlorate,perchloric acid, cesium salt 1:1,clo4.cs,acmc-20alh8,caesium 1+ perchlorate ion
123309
cesium;perchlorate
[O-]Cl(=O)(=O)=O.[Cs+]
13454-84-7
ClCsO4
232.351
MFCD00010956
WKDKOOITVYKILI-UHFFFAOYSA-M
cesium perchlorate,caesium 1+ ion perchlorate,perchloric acid, cesium salt,caesiumperchlorat,caesium perchlorate,perchloric acid, cesium salt 1:1,clo4.cs,acmc-20alh8,caesium 1+ perchlorate ion
123309
cesium;perchlorate
[O-]Cl(=O)(=O)=O.[Cs+]
Oxidation reagent
Assay Percent Range 99%
Linear Formula CsClO4
Formula Weight 232.36
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Packaging Glass Bottle
Solubility Solubility in water: 1.9g/100mL.
Color White
Melting Point 250.0°C
Quantity 10g
Chemical Name or Material Cesium perchlorate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger