Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium, plasma standard solution, Cs 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13825AD

Quantity 50mL

 View more versions of this product

Catalog No. AA13825AD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7697-37-2,534-17-8,7732-18-5
Cs2CO3 in 5% HNO3
Cesium
Liquid
MFCD00134037

Specifications

7697-37-2,534-17-8,7732-18-5
Liquid
MFCD00134037
23 hPa (17mm Hg) at 20°C
50mL
Clear liquid
Cs2CO3 in 5% HNO3
Nitric acid: 5.0%; Cesium carbonate: 0.122%; Water: 94.878%
Clear colorless
Cesium

Safety and Handling

SDS & Safety Documents

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit