Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Sulfate, 99.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
10294-54-9
10294-54-9
Grade Reagent
CAS Max % 1
Percent Purity 0.995
Heavy Metals 2e-05
Iron (Fe) 2e-05
Lithium (Li) 2e-05
Magnesium (Mg) 5e-05
Packaging Amber Glass Bottle
Potassium (K) 0.0005
Rubidium (Rb) 0.002
Sodium (Na) 0.002
Quantity 25 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.