Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Sulfate, 99.9% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
25g
100g
10294-54-9
Cs2O4S
361.867
MFCD00010959
FLJPGEWQYJVDPF-UHFFFAOYSA-L
cesium sulfate,dicesium sulfate,sulfuric acid, dicesium salt,caesium sulphate,caesium sulfate,unii-8d6r91cs62,dicaesium 1+ ion sulfate,cesiumsulfate,dicaesium 1+ sulfate
25137
dicesium;sulfate
[O-]S(=O)(=O)[O-].[Cs+].[Cs+]
10294-54-9
Cs2O4S
361.867
MFCD00010959
FLJPGEWQYJVDPF-UHFFFAOYSA-L
cesium sulfate,dicesium sulfate,sulfuric acid, dicesium salt,caesium sulphate,caesium sulfate,unii-8d6r91cs62,dicaesium 1+ ion sulfate,cesiumsulfate,dicaesium 1+ sulfate
25137
dicesium;sulfate
[O-]S(=O)(=O)[O-].[Cs+].[Cs+]
Assay Percent Range 99.9% (Metal basis)
Appearance White powder
Solubility Information Very soluble in water. Slightly soluble in alcohol,acetone,pyridine.
Formula Weight 361.87
Physical Form -20 Mesh Powder
Density 4.243
Sensitivity Hygroscopic
Color White
Melting Point 1010°C
Quantity 100g
Chemical Name or Material Cesium sulfate
Show More Show Less

P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H361f-H373

EINECSNumber : 233-662-6

TSCA : Yes

Recommended Storage : Ambient temperatures