Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cesium sulfate, Puratronic™, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 1111209

Quantity 10g

 View more versions of this product

Catalog No. AA1111209


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10294-54-9
361.87
14,2016
Puratronic™
Cs2SO4
MFCD00010959
Cesium sulfate

Specifications

10294-54-9
Cs2SO4
MFCD00010959
4.243g/mL
Hygroscopic
1,010°C
Cesium sulfate
Puratronic™
Powder
361.87
14,2016
Odorless
White
10g
99.997%, (metals basis)

Safety and Handling

SDS & Safety Documents

RTECSNumber : FL0800000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit