Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Sulfate, Ultra High Purity, 99.9%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
10294-54-9
10294-54-9
Grade High Purity
CAS Max % 1
Percent Purity 0.999
Chromium (Cr) 1e-06
Copper (Cu) 1e-06
Lead (Pb) 1e-06
Lithium (Li) 1e-05
Magnesium (Mg) 1e-06
Nickel (Ni) 5e-06
Packaging Amber Glass Bottle
Potassium (K) 5e-05
Rubidium (Rb) 0.0002
Sodium (Na) 0.0001
Quantity 25 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.