Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Trifluoroacetate (CsTFA), illustra™, GE Healthcare

Click to view available options
Quantity:
100mL
CsTFA, aqueous solution
Cesium Trifluoroacetate Solution
CsTFA, aqueous solution
Cesium Trifluoroacetate Solution

  • Effective inhibitor of ribonucleases
  • Can separate RNA and DNA
  • Soluble in ethanol so nucleic acids can be recovered from the gradient by ethanol precipitation

For the separation and purification of nucleic acids, especially RNA, by isopycnic centrifugation
Density 2.0 ± 0.05 g/mL
Quantity 100mL