Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cesium triiodide, 98%

Click to view available options
Quantity:
1g
20202-54-4
CsI3-2
513.619
MFCD00016053
UBARFHYEKZLYQX-UHFFFAOYSA-K
cesiumtriiodide
5810786
cesium;triiodide
[I-].[I-].[I-].[Cs+]
20202-54-4
CsI3-2
513.619
MFCD00016053
UBARFHYEKZLYQX-UHFFFAOYSA-K
cesiumtriiodide
5810786
cesium;triiodide
[I-].[I-].[I-].[Cs+]
Formula Weight 513.62
Percent Purity 98%
Quantity 1g
Chemical Name or Material Cesium triiodide

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning